missing translation for 'onlineSavingsMsg'
Learn More

Cocaine Hydrochloride Solution, 1.0 mg/mL in Methanol, Analytical Standard, For Drug Analysis, MilliporeSigma™ Supelco™

Catalog No. 111017176
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017176 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017176 Supplier Merck Emd Millipore Supplier No. C15281ML
Only null left

Cocaine is mostly commonly available as cocaine hydrochloride which is a white to off-white powder. It is taken by intranasal route. In some surgeries it is used as topical anaesthetic drug.

Specifications

Packaging Ampule
CAS 53-21-4
Concentration 1.0 mg/mL in methanol
Linear Formula C17H21NO4 · HCl
MDL Number MFCD00058119
Industry Type Drug Analysis
Quantity 1 mL
Formula Weight 339.81
Grade Analytical Standard