missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt Sulfate, 7-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor J.T.Baker 169605
Specifications
| Cobalt Sulfate | |
| 10026-24-1 | |
| 2.5 kg | |
| MFCD00149658 | |
| MEYVLGVRTYSQHI-UHFFFAOYSA-L | |
| λ²-cobalt(2+) heptahydrate sulfate | |
| 61444 | |
| BAKER ANALYZED™ Reagent | |
| 1L = 2.03kg |
| 7-Hydrate | |
| Crystals | |
| CoH14O11S | |
| cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o | |
| O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O | |
| 281.09 | |
| 281.1 | |
| Poly Bottle |
Chemical Identifiers
| 10026-24-1 | |
| 281.09 | |
| MEYVLGVRTYSQHI-UHFFFAOYSA-L | |
| 61444 | |
| O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O |
| CoH14O11S | |
| MFCD00149658 | |
| cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o | |
| λ²-cobalt(2+) heptahydrate sulfate |