missing translation for 'onlineSavingsMsg'
Learn More

Cobalt naphthenate, Co 6%, approximately 53% in mineral spirits

Catalog No. AA40387A3
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40387A3 2 kg
AA4038736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40387A3 Supplier Thermo Scientific Chemicals Supplier No. 040387A3

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61789-51-3 | C20H34CoO4 | 397.42 g/mol

Cobalt naphthenate is widely used in polyester resins and paint driers. It is used as a curing accelerator, cross linker catalyst for unsaturated polyester resins. It is also used in the production of adhesives, varnishes and waterproofing agents for textiles. Further, it is used as corrosion inhibitors, lubricants, and fuel additives. It plays an important role as oil drying agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61789-51-3
Molecular Formula C20H34CoO4
Molecular Weight (g/mol) 397.42
MDL Number MFCD02094763
InChI Key XSWKLHINRKWMTD-UHFFFAOYNA-L
Synonym cobalt naphthenate, cobalt ii 2-naphthoate
PubChem CID 119077305
IUPAC Name cobalt;naphthalene-2-carboxylic acid
SMILES [Co++].CCC1CCC(CCC([O-])=O)C1.CCC1CCC(CCC([O-])=O)C1

Specifications

Flash Point 40°C (104°F)
Quantity 2 kg
UN Number UN1993
Solubility Information Insoluble in water.
Concentration or Composition (by Analyte or Components) Co 6%, approximately 53% in min.eral spirits
Physical Form Liquid
Chemical Name or Material Cobalt naphthenate
Hazard Category H226-H302-H304-H317-H336-H350-H372
Hazard Statement GHS H Statement
H304-H351-H226-H317
May be fatal if swallowed and enters airways.
Suspected of causing cancer.
Flammable liquid and vapor.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280g-P281-P301+P310-P303+P361+P353-P304+P340-P308+P313-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 263-064-0
RTECSNumber QK8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only