missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) tetrafluoroborate hydrate, 96%

CAS: 26490-63-1

$111.22 - $324.82

Chemical Identifiers

CAS 26490-63-1
MDL Number MFCD00016012
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA2314214
View Documents
Thermo Scientific Chemicals
02314214
25 g
Each for $111.22
Only null left
 
AA2314222
View Documents
Thermo Scientific Chemicals
02314222
100 g
Each for $324.82
Only null left
 
Description

Description

It is a cobalt salt. It is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

26490-63-1
MFCD00016012

Specifications

26490-63-1
25 g
UN3260
232.54 (Anhydrous)
Hygroscopic
Crystalline
MFCD00016012
Soluble in water.
96%
Cobalt(II) tetrafluoroborate hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H341-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P271-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H334-H335-H341-H350

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only