accessibility menu, dialog, popup

UserName

CoFBFFFFBFFF

CAS RN 26490-63-1

IUPAC Name: λ²-cobalt(2+) bis(tetrafluoroboranuide)
Synonyms: λ²-cobalt(2+) ditetrafluoroborate
Molecular Weight (g/mol): 232.54
Molecular Formula: B2CoF8
InChi Key: SSIMDEJNGHHZCA-UHFFFAOYSA-N
SMILES: [Co++].F[B-](F)(F)F.F[B-](F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results