Learn More
Cobalt(II) sulfate hydrate, Reagent, Co 20.8% min
Ceramics, pigments, glazes, plating baths, catalyst, storage batteries, paint/ink drier | CAS: 60459-08-7 | CoO4S | 154.99 g/mol
$173.71 - $598.44
Chemical Identifiers
| CAS | 60459-08-7 |
|---|---|
| Molecular Formula | CoO4S |
| Molecular Weight (g/mol) | 154.99 |
| MDL Number | MFCD00149657 |
| InChI Key | KTVIXTQDYHMGHF-UHFFFAOYSA-L |
| Synonym | cobalt ii sulfate hydrate, cobalt sulfate monohydrate, unii-5zqa69klc0, 5zqa69klc0, cobalt mesosulfate coh2so5, cobalt sulfate coso4 monohydrate, cobalt ii sulfatehydrate, cobalt sulphate monohydrate, co.so4.h2o, cobalt 2+ hydrate sulfate |
| PubChem CID | 16210984 |
| IUPAC Name | cobalt(2+);sulfate;hydrate |
| SMILES | [Co++].[O-]S([O-])(=O)=O |
Description
Ceramics, pigments, glazes, plating baths, catalyst, storage batteries, paint/ink drier
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 60459-08-7 | |
| 154.99 | |
| KTVIXTQDYHMGHF-UHFFFAOYSA-L | |
| 16210984 | |
| [Co++].[O-]S([O-])(=O)=O |
| CoO4S | |
| MFCD00149657 | |
| cobalt ii sulfate hydrate, cobalt sulfate monohydrate, unii-5zqa69klc0, 5zqa69klc0, cobalt mesosulfate coh2so5, cobalt sulfate coso4 monohydrate, cobalt ii sulfatehydrate, cobalt sulphate monohydrate, co.so4.h2o, cobalt 2+ hydrate sulfate | |
| cobalt(2+);sulfate;hydrate |
Specifications
| 60459-08-7 | |
| Crystalline | |
| CoO4S | |
| MFCD00149657 | |
| cobalt ii sulfate hydrate, cobalt sulfate monohydrate, unii-5zqa69klc0, 5zqa69klc0, cobalt mesosulfate coh2so5, cobalt sulfate coso4 monohydrate, cobalt ii sulfatehydrate, cobalt sulphate monohydrate, co.so4.h2o, cobalt 2+ hydrate sulfate | |
| KTVIXTQDYHMGHF-UHFFFAOYSA-L | |
| cobalt(2+);sulfate;hydrate | |
| 16210984 | |
| Reagent | |
| Cobalt(II) sulfate hydrate |
| Red to Orange | |
| 250 g | |
| CoSO4·xH2O | |
| UN3077 | |
| Soluble in water; Slightly soluble in methanol and ethanol | |
| [Co++].[O-]S([O-])(=O)=O | |
| 154.99 | |
| 154.99 (Anhydrous) | |
| Co 20.8% min. |
Safety and Handling
GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P272-P280-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c
H302-H317-H334-H341-H350-H360-H360F-H500
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 233-334-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only