missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) sulfate hydrate, Reagent, Co 20.8% min

Ceramics, pigments, glazes, plating baths, catalyst, storage batteries, paint/ink drier | CAS: 60459-08-7 | CoO4S | 154.99 g/mol

Supplier:  Thermo Scientific Chemicals 01134030

 View more versions of this product

Catalog No. AA1134030

Only null left
Explore available promotions

Description

Description

Ceramics, pigments, glazes, plating baths, catalyst, storage batteries, paint/ink drier

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cobalt(II) sulfate hydrate
Red to Orange
250 g
CoSO4·xH2O
UN3077
Soluble in water; Slightly soluble in methanol and ethanol
[Co++].[O-]S([O-])(=O)=O
154.99
154.99 (Anhydrous)
Co 20.8% min.
60459-08-7
Crystalline
CoO4S
MFCD00149657
cobalt ii sulfate hydrate, cobalt sulfate monohydrate, unii-5zqa69klc0, 5zqa69klc0, cobalt mesosulfate coh2so5, cobalt sulfate coso4 monohydrate, cobalt ii sulfatehydrate, cobalt sulphate monohydrate, co.so4.h2o, cobalt 2+ hydrate sulfate
KTVIXTQDYHMGHF-UHFFFAOYSA-L
cobalt(2+);sulfate;hydrate
16210984
Reagent

Chemical Identifiers

60459-08-7
154.99
KTVIXTQDYHMGHF-UHFFFAOYSA-L
16210984
[Co++].[O-]S([O-])(=O)=O
CoO4S
MFCD00149657
cobalt ii sulfate hydrate, cobalt sulfate monohydrate, unii-5zqa69klc0, 5zqa69klc0, cobalt mesosulfate coh2so5, cobalt sulfate coso4 monohydrate, cobalt ii sulfatehydrate, cobalt sulphate monohydrate, co.so4.h2o, cobalt 2+ hydrate sulfate
cobalt(2+);sulfate;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P272-P280-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c

H302-H317-H334-H341-H350-H360-H360F-H500

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 233-334-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only