missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis)

Catalog No. p-7048499
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
AA1069614 25 g
AA1069622 100 g
2 options

Catalog No. AA1069614

Supplier: Thermo Scientific Chemicals 01069614

CAS: 10026-24-1 | CoH14O11S | 281.09 g/mol

Cobalt(II) sulfate heptahydrate is used in the preparation of pigments and other cobalt salts. It is used in storage batteries and electroplating baths, sympathetic inks, and as an additive to soils and animal feeds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10026-24-1
Molecular Formula CoH14O11S
Molecular Weight (g/mol) 281.09
MDL Number MFCD00149658
InChI Key MEYVLGVRTYSQHI-UHFFFAOYSA-L
Synonym cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o
PubChem CID 61444
SMILES O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O

Specifications

Color Red
pH 4
Physical Form Crystalline Aggregates
Quantity 25 g
Linear Formula CoSO4·7H2O
UN Number UN3077
Merck Index 14,2448
Solubility Information Soluble in water.
IUPAC Name cobalt(2+);sulfate;heptahydrate
Formula Weight 281.10 (154.99 Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Density 2.03 g/mL
Assay (metals basis)
Chemical Name or Material Cobalt(II) sulfate heptahydrate
Show More Show Less
Hazard Category H302-H317-H334-H341-H350i-H360F
Hazard Statement GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 233-334-2
RTECSNumber GG3200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only