missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) sulfate heptahydrate, for analysis

Catalog No. p-7077446
Change view
Click to view available options
Quantity:
250 g
1 kg
Catalog No. Quantity
AC453902500 250 g
AC453900010 1 kg
2 options

Catalog No. AC453902500

Supplier: Thermo Scientific Chemicals 453902500

CAS: 10026-24-1 | CoH14O11S | 281.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10026-24-1
Molecular Formula CoH14O11S
Molecular Weight (g/mol) 281.09
MDL Number MFCD00149658
InChI Key MEYVLGVRTYSQHI-UHFFFAOYSA-L
Synonym cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o
PubChem CID 61444
SMILES O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O

Specifications

Melting Point 98.0°C
pH 4 to 6 (5% aq. soln.)
Quantity 250 g
Linear Formula CoSO4·7H2O
Merck Index 15,2432
Solubility Information Solubility in water: 362g/L (20°C). Other solubilities: slightly soluble in methanol and ethanol
IUPAC Name cobalt(2+);sulfate;heptahydrate
Formula Weight 281.09
Percent Purity 99.0 to 101.0% (Complexometry)
Grade Analytical
Chemical Name or Material Cobalt(II) sulfate heptahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENT

RUO – Research Use Only