missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) sulfate heptahydrate, 99+%, extra pure

Catalog No. p-222888
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC213101000 100 g Glass bottle
AC213105000 500 g Glass bottle
AC213100025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC213101000 Supplier Thermo Scientific Chemicals Supplier No. 213101000
Only null left

CAS: 10026-24-1 | CoH14O11S | 281.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 10026-24-1
Molecular Formula CoH14O11S
Molecular Weight (g/mol) 281.09
MDL Number MFCD00149658
InChI Key MEYVLGVRTYSQHI-UHFFFAOYSA-L
Synonym cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o
PubChem CID 61444
SMILES O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O

Specifications

Melting Point 98.0°C
Color Red to Brown
Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula CoSO4·7H2O
Merck Index 15,2432
Solubility Information Solubility in water: 362g/L (20°C). Other solubilities: slightly soluble in methanol and ethanol
IUPAC Name λ²-cobalt(2+) heptahydrate sulfate
Formula Weight 281.09
Percent Purity ≥99%
Grade Extra Pure
Packaging Glass bottle
Density 2.03
Chemical Name or Material Cobalt(II) sulfate heptahydrate, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENT
RTECSNumber GG3200000

RUO – Research Use Only