missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) sulfamate hydrate

CAS: 14017-41-5 | CoH4N2O6S2 | 251.09 g/mol

$243.62 - $2206.63

Chemical Identifiers

CAS 14017-41-5
Molecular Formula CoH4N2O6S2
Molecular Weight (g/mol) 251.09
MDL Number MFCD00049877
InChI Key WLQXLCXXAPYDIU-UHFFFAOYSA-L
Synonym cobaltous sulfamate, cobalt sulfamate, cobalt ii sulfamate, cobalt 2+ disulfamate, unii-0uhd1pcy6u, 0uhd1pcy6u, cobalt 2+ disulphamate, cobaltous disulfamate, acmc-20alek
PubChem CID 26398
SMILES [Co++].NS([O-])(=O)=O.NS([O-])(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4390622
View Documents
Thermo Scientific Chemicals
04390622
100 g
Each for $243.62
Only null left
 
AA4390636
View Documents
Thermo Scientific Chemicals
04390636
500 g
Each for $744.96
Only null left
 
AA43906A3
View Documents
Thermo Scientific Chemicals
043906A3
2 kg
Each for $2,206.63
Only null left
 
Description

Description

Cobalt(II) sulfamate hydrate is used in photonic device fabrication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14017-41-5
251.09
WLQXLCXXAPYDIU-UHFFFAOYSA-L
26398
CoH4N2O6S2
MFCD00049877
cobaltous sulfamate, cobalt sulfamate, cobalt ii sulfamate, cobalt 2+ disulfamate, unii-0uhd1pcy6u, 0uhd1pcy6u, cobalt 2+ disulphamate, cobaltous disulfamate, acmc-20alek
[Co++].NS([O-])(=O)=O.NS([O-])(=O)=O

Specifications

14017-41-5
100 g
MFCD00049877
Soluble in water.
[Co++].NS([O-])(=O)=O.NS([O-])(=O)=O
251.09
251.11 (Anhydrous)
Cobalt (II) sulfamate hydrate
Powder
CoH4N2O6S2
cobaltous sulfamate, cobalt sulfamate, cobalt ii sulfamate, cobalt 2+ disulfamate, unii-0uhd1pcy6u, 0uhd1pcy6u, cobalt 2+ disulphamate, cobaltous disulfamate, acmc-20alek
WLQXLCXXAPYDIU-UHFFFAOYSA-L
λ²-cobalt(2+) disulfamate
26398
Odorless
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P272-P280-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c

H302-H315-H317-H319-H341-H350i-H360F

EINECSNumber : 237-834-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only