missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) sulfamate hydrate

CAS: 14017-41-5 | CoH4N2O6S2 | 251.09 g/mol

Supplier:  Thermo Scientific Chemicals 04390622

 View more versions of this product

Catalog No. AA4390622


Only null left
Explore available promotions

Description

Description

Cobalt(II) sulfamate hydrate is used in photonic device fabrication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cobalt (II) sulfamate hydrate
Powder
CoH4N2O6S2
cobaltous sulfamate, cobalt sulfamate, cobalt ii sulfamate, cobalt 2+ disulfamate, unii-0uhd1pcy6u, 0uhd1pcy6u, cobalt 2+ disulphamate, cobaltous disulfamate, acmc-20alek
WLQXLCXXAPYDIU-UHFFFAOYSA-L
λ²-cobalt(2+) disulfamate
26398
Odorless
14017-41-5
100 g
MFCD00049877
Soluble in water.
[Co++].NS([O-])(=O)=O.NS([O-])(=O)=O
251.09
251.11 (Anhydrous)

Chemical Identifiers

14017-41-5
251.09
WLQXLCXXAPYDIU-UHFFFAOYSA-L
26398
CoH4N2O6S2
MFCD00049877
cobaltous sulfamate, cobalt sulfamate, cobalt ii sulfamate, cobalt 2+ disulfamate, unii-0uhd1pcy6u, 0uhd1pcy6u, cobalt 2+ disulphamate, cobaltous disulfamate, acmc-20alek
[Co++].NS([O-])(=O)=O.NS([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P272-P280-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c

H302-H315-H317-H319-H341-H350i-H360F

EINECSNumber : 237-834-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only