missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Phthalocyanine 93.0+%, TCI America™

Supplier:  TCI America P08875G

 View more versions of this product

Catalog No. P08875G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Cobalt(II) Phthalocyanine
300°C
C32H16CoN8
5 g
MPMSMUBQXQALQI-UHFFFAOYSA-N
571.469
571.47
Crystalline Powder
3317-67-7
Purple-Red
MFCD00010718
cobalt ii phthalocyanine, cobalt phthalocyanine, phthalocyanine cobalt ii, phthalocyanine, cobalt, cobalt ii phthalocyanine, beta-form, dye content 97 %
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Co+2]
2734991
≥93.0% (T)

Chemical Identifiers

3317-67-7
571.469
MPMSMUBQXQALQI-UHFFFAOYSA-N
2734991
C32H16CoN8
MFCD00010718
cobalt ii phthalocyanine, cobalt phthalocyanine, phthalocyanine cobalt ii, phthalocyanine, cobalt, cobalt ii phthalocyanine, beta-form, dye content 97 %
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Co+2]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-4131

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions