missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Phthalocyanine 93.0+%, TCI America™

Catalog No. p-7134829
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08875G 5 g
P088725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08875G Supplier TCI America Supplier No. P08875G
Only null left

Chemical Identifiers

CAS 3317-67-7
Molecular Formula C32H16CoN8
Molecular Weight (g/mol) 571.469
MDL Number MFCD00010718
InChI Key MPMSMUBQXQALQI-UHFFFAOYSA-N
Synonym cobalt ii phthalocyanine, cobalt phthalocyanine, phthalocyanine cobalt ii, phthalocyanine, cobalt, cobalt ii phthalocyanine, beta-form, dye content 97 %
PubChem CID 2734991
SMILES C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Co+2]

Specifications

Melting Point 300°C
Color Purple-Red
Quantity 5 g
Formula Weight 571.47
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cobalt(II) Phthalocyanine
EINECSNumber (5)-4131
TSCA Yes