missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) oxalate dihydrate, Reagent Grade

Catalog No. p-7080379
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8775806 5 g
AA8775814 25 g
AA8775822 100 g
AA8775836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA8775806 Supplier Thermo Scientific Chemicals Supplier No. 08775806
Only null left

Stabilizer for HCN, temperature indicator | CAS: 5965-38-8 | C2H4CoO6 | 182.981 g/mol

Cobalt(II) oxalate dihydrate is used as a stabilizer for hydrogen cyanide and as a temperature indicator. It is used in the preparation of cobalt catalysts, and cobalt metal powder for powder-metallurgical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5965-38-8
Molecular Formula C2H4CoO6
Molecular Weight (g/mol) 182.98
MDL Number MFCD03414148
InChI Key MWHSMSAKVHVSAS-UHFFFAOYSA-L
Synonym cobaltousoxalate,cobaltoxalate
PubChem CID 131881712
IUPAC Name 1,3-dioxa-2-cobaltacyclopentane-4,5-dione dihydrate
SMILES O.O.O=C1O[Co]OC1=O

Specifications

Melting Point 229°C to 231°C (decomposition)
Density 3.021 g/mL
Odor Odorless
Quantity 5 g
UN Number UN3288
Merck Index 14,2445
Solubility Information Soluble in aqueous ammonia; Insoluble in water; Slightly soluble in acids.
Formula Weight 182.98 (146.95 Anhydrous)
Grade Reagent
Physical Form Powder
Chemical Name or Material Cobalt (II) oxalate dihydrate
Show More Show Less
Hazard Category H302+H312-H350
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P302+P352-P308+P313-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 212-409-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only