accessibility menu, dialog, popup

UserName

CAS RN 5965-38-8

IUPAC Name: 1,3-dioxa-2-cobaltacyclopentane-4,5-dione dihydrate
Synonyms: cobaltous oxalate dihydrate
Molecular Weight (g/mol): 182.98
Molecular Formula: C2H4CoO6
InChi Key: MWHSMSAKVHVSAS-UHFFFAOYSA-L
SMILES: O.O.O=C1O[Co]OC1=O
Grade 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (4)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)