missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) isopropoxide

Catalog No. AA4281006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4281006 5 g
AA4281003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4281006 Supplier Thermo Scientific Chemicals Supplier No. 04281006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60406-95-3 | C6H14CoO2 | 177.109 g/mol

Cobalt(II) isopropoxide is used in display & imaging, catalysis & synthesis and ferroelectric metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon

Chemical Identifiers

CAS 60406-95-3
Molecular Formula C6H14CoO2
Molecular Weight (g/mol) 177.109
MDL Number MFCD00799075
InChI Key VBXHMSMVQZJJLD-UHFFFAOYSA-N
Synonym cobalt ii isopropoxide, cobalt isopropoxide, acmc-20alem, cobalt ii diisopropoxide, cobalt 2+ dipropan-2-olate, 2-propanol, cobalt 2+ salt
PubChem CID 18517386
IUPAC Name cobalt(2+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].[Co+2]

Specifications

Packaging Ampule
Quantity 5 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 178.11
Physical Form Powder
Chemical Name or Material Cobalt (II) isopropoxide
Hazard Category H228-H315-H319-H335-H341
Hazard Statement GHS H Statement
H228-H341-H315-H319-H335
Flammable solid.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P240-P241-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA No
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only