missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chloride hexahydrate, pure

Catalog No. p-7077193
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC192862500 250 g
AC192860010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC192862500 Supplier Thermo Scientific Chemicals Supplier No. 192862500
Only null left

CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Melting Point 86.0°C
Quantity 250 g
Linear Formula CoCl2·6H2O
Fieser 01,155; 06,127; 11,479
Merck Index 15,2421
Solubility Information Solubility in water: 970g/L (20°C).
Formula Weight 237.93
Percent Purity 98.0 to 102.0% (Complexometry)
Grade Pure
Chemical Name or Material Cobalt(II) chloride hexahydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage fertility.
Harmful if inhaled.
Harmful if swallowed.
Suspected of causing genetic defects.
May cause cancer by inhalation.
May cause an allergic skin reaction.
May cause allergy or ast
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:

RUO – Research Use Only