missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chloride hexahydrate, Puratronic™, 99.998% (metals basis)

Catalog No. AA1069209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1069209 10 g
AA1069218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1069209 Supplier Thermo Scientific Chemicals Supplier No. 01069209

CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol

Cobalt(II) chloride hexahydrate has been used to study apoptotic effects in HepG2 cells. Also reported as a stabilizer for reconstitution of the radiopharmaceutical technetium-99m exametazime. It has been used in studies to examine heat shock protein expression of inflamed skin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Melting Point 87°C
Color Purple-Red
pH 4.9
Physical Form Crystalline Aggregates
Quantity 10 g
Linear Formula CoCl2·6H2O
UN Number UN3077
Merck Index 14,2437
Solubility Information Soluble in water,acetone,ether,and alcohols.
Formula Weight 237.93 (129.84 Anhydrous)
Percent Purity 99.998%
Grade Puratronic
Odor Odorless
Sensitivity Hygroscopic
Density 1.924 g/mL
Assay (metals basis)
Chemical Name or Material Cobalt(II) chloride hexahydrate
Show More Show Less
Hazard Category H302+H332-H317-H334-H341-H350i-H360F
Hazard Statement GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 231-589-4
RTECSNumber GG0200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only