missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(Ii) Chloride Hexahydrate, ACS Reagent, Honeywell Fluka™

Catalog No. p-7071943
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000420 5 g
6000418 100 g
6000419 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6000420 Supplier Honeywell Chemicals Supplier No. 2555995G
Only null left

ACS Reagent, 98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Quantity 5 g
Linear Formula CoCl2 · 6H2O
UN Number 3077
Formula Weight 237.93g/mol
Percent Purity 98%
Grade ACS Reagent
Packaging Glass Bottle
Vapor Pressure 40mmHg (0°C)
Chemical Name or Material Cobalt(Ii) Chloride Hexahydrate
Hazard Statement P201-P261-P273-P280-P284-P304 + P340
Precautionary Statement H302-H317-H334-H341-H350i-H360F-H410
EINECSNumber 231-589-4
RTECSNumber GG0200000