missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chloride hexahydrate, 99.9% (metals basis)

Catalog No. AA1134430
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1134430 250 g
AA11344A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1134430 Supplier Thermo Scientific Chemicals Supplier No. 01134430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Sensitivity: Hygroscopic; m.p. 52-56°C -4H2O, 100°C -5H2O, 120-140°C -6H2O | CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol

Cobalt(II) chloride hexahydrate is used as a humidity indicator in weather instruments. It finds use in electroplating of cobalt, in invisible ink, in organic chemistry and is a precursor to cobaltocene(bis (cyclopentadienyl) cobalt(II)).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Melting Point 87°C
Color Purple-Red
Physical Form Crystals
Quantity 250 g
Assay Percent Range 99.9% (Metals basis)
Solubility Information Soluble in water,alcohol,acetone,ether and glycerol.
Formula Weight 237.93 (129.84 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Density 1.924
Chemical Name or Material Cobalt(II) chloride hexahydrate
Show More Show Less
Hazard Category H302+H332-H317-H334-H341-H350i-H360F
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 231-589-4
RTECSNumber GG0200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only