missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) benzoate

Catalog No. p-7080370
missing translation for 'changeView'
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8738009 10 g
AA8738018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AA8738009 Supplier Thermo Scientific Chemicals missing translation for 'supplierNo' 08738009
Only null left

CAS: 932-69-4 | C14H10CoO4 | 301.16 g/mol

Cobalt(II) benzoate is used as ingredients of many cosmetic products, industrial fields such as automobile, fuel, battery, paint, metallurgy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 932-69-4
Molecular Formula C14H10CoO4
Molecular Weight (g/mol) 301.16
MDL Number MFCD00050769
InChI Key GAYAMOAYBXKUII-UHFFFAOYSA-L
Synonym cobalt 2+ dibenzoate, cobalt ii benzoate, cobalt dibenzoate, cobalt benzoate, benzoic acid, cobalt 2+ salt, benzoic acid, cobalt 2+ salt 2:1, cobaltous benzoate, dibenzoic acid cobalt ii salt
PubChem CID 164760
IUPAC Name cobalt(2+);dibenzoate
SMILES [Co++].[O-]C(=O)C1=CC=CC=C1.[O-]C(=O)C1=CC=CC=C1

Specifications

Melting Point 114°C to 116°C
Color Gray
Odor Odorless
Linear Formula Co(OOCC6H5)2
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 301.16
Physical Form Crystalline
Chemical Name or Material Cobalt (II) benzoate
Hazard Category H334-H350
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P281-P285-P304+P341-P308+P313-P342+P311-P501c
EINECSNumber 213-257-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only