missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) 2,4-pentanedionate

Catalog No. p-7049029
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1253718 50 g
AA1253730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1253718 Supplier Thermo Scientific Chemicals Supplier No. 01253718
Only null left

CAS: 14024-48-7 | C10H14CoO4 | 257.15 g/mol

Cobalt(II) 2,4-pentanedionate is used as an important catalyst for organic reactions. It is also used as paint dryer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14024-48-7
Molecular Formula C10H14CoO4
Molecular Weight (g/mol) 257.15
MDL Number MFCD00000014
InChI Key UTYYEGLZLFAFDI-FDGPNNRMSA-L
Synonym Cobalt(II) acetylacetonate; Bis(acetylacetonato)cobalt(II)
IUPAC Name λ²-cobalt(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Co++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 165°C to 170°C
Color Blue-Purple
Odor Odorless
Quantity 50 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 257.14
Physical Form Crystalline
Chemical Name or Material Cobalt(II) 2,4-pentanedionate
Hazard Category H302-H317-H318-H334-H341-H350
Hazard Statement GHS H Statement
H334-H302-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P305+P351+P338-P308+P313-P310-P330-P333+P313-P342+P311-P363-P501c
EINECSNumber 237-855-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only