accessibility menu, dialog, popup

UserName

CAS RN 14024-48-7

OOOOH3CH3CCH3CH3Co

CAS RN 14024-48-7

IUPAC Name: λ²-cobalt(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Synonyms: λ²-cobalt(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 257.15
Molecular Formula: C10H14CoO4
InChi Key: UTYYEGLZLFAFDI-FDGPNNRMSA-L
SMILES: [Co++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Physical Form 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)