missing translation for 'onlineSavingsMsg'
Learn More

Cobalt Chloride Hexahydrate, Purified, Reagents

Catalog No. 01337311
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337311 100 g
01337310 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337311 Supplier Labchem Inc Supplier No. C2181000100C2
Only null left

Cobalt Chloride Hexahydrate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
Synonym Cobalt(II) Chloride Hexahydrate, Cobaltous Chloride Hexahydrate
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Boiling Point 1049°C
Melting Point 56°C
Color Pink to Red
pH 4.9
Physical Form Solid
Quantity 100 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 1.924
Chemical Name or Material Cobalt Chloride Hexahydrate, Purified