Learn More
Cobalt, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 453910010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Cobalt | |
| 1kg | |
| MFCD00010935 | |
| Solubility in water: insoluble. | |
| [Co] | |
| 58.93 | |
| CHEBI:27638 | |
| ≥99% |
| 7440-48-4 | |
| Co | |
| kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride | |
| GUTLYIVDDKVIGB-UHFFFAOYSA-N | |
| cobalt | |
| 104730 | |
| 58.93 | |
| Pure |
Chemical Identifiers
| 7440-48-4 | |
| 58.93 | |
| GUTLYIVDDKVIGB-UHFFFAOYSA-N | |
| 104730 | |
| cobalt |
| Co | |
| MFCD00010935 | |
| kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride | |
| CHEBI:27638 | |
| [Co] |
Safety and Handling
GHS H Statement Flammable solid. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if swallowed. Fatal if inhaled. Causes serious eye irritation. May cause cancer by inhalation. May damage fertility. Very toxic to aquatic life with long lasting effects.
GHS P Statement Use personal protective equipment as required. Obtain special instructions before use. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Immediately call a POISON CENTER or doctor/physician.
Danger
RUO â Research Use Only