missing translation for 'onlineSavingsMsg'
Learn More

Cobalt, 99+%, Thermo Scientific™

Numéro de catalogue. p-7077447
Modifier l'affichage
Click to view available options
Quantité:
250g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
AC453912500 250g
AC453910010 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC453912500 Fournisseur Thermo Scientific Chemicals Code fournisseur 453912500

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 7440-48-4
Formule moléculaire Co
Poids moléculaire (g/mol) 58.93
Numéro MDL MFCD00010935
Clé InChI GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonyme kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
Nom de l’IUPAC cobalt
SOURIRES [Co]

Spécifications

Nom ou substance chimique Cobalt
Quantité 250g
Renseignements sur la solubilité Solubility in water: insoluble.
Poids de formule 58.93
Pourcentage de pureté ≥99%
Niveau Pure
Hazard Category Danger
Hazard Statement GHS H Statement Flammable solid. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if swallowed. Fatal if inhaled. Causes serious eye irritation. May cause cancer by inhalation. May damage fertility. Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement Use personal protective equipment as required. Obtain special instructions before use. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only