missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CNTNAP1 Recombinant Rabbit Monoclonal Antibody (23GB5605)
GREENER_CHOICE

Catalog No. MA553910
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553910 100 μL
MA553911 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553910 Supplier Invitrogen™ Supplier No. MA553910
Only null left

Rabbit Recombinant Monoclonal Antibody

CNTNAP1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

CNTNAP1 was initially identified as a 190-kDa protein associated with the contactin-PTPRZ1 complex. The 1, 384-amino acid protein, also designated p190 or CASPR for 'contactin-associated protein, ' includes an extracellular domain with several putative protein-protein interaction domains, a putative transmembrane domain, and a 74-amino acid cytoplasmic domain. The architecture of its extracellular domain is similar to that of neurexins, and this protein may be the signaling subunit of contactin, enabling recruitment and activation of intracellular signaling pathways in neurons. The gene product was initially identified as a 190-kDa protein associated with the contactin-PTPRZ1 complex. The 1, 381-amino acid protein, also designated p190 or CASPR for 'contactin-associated protein, ' includes an extracellular domain with several putative protein-protein interaction domains, a putative transmembrane domain, and a 74-amino acid cytoplasmic domain. Northern blot analysis showed that the gene is transcribed predominantly in brain as a transcript of 6.2 kb, with weak expression in several other tissues tested. The architecture of its extracellular domain is similar to that of neurexins, and this protein may be the signaling subunit of contactin, enabling recruitment and activation of intracellular signaling pathways in neurons.

Specifications

Antigen CNTNAP1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5605
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cntnap1
Gene Accession No. O54991, P78357
Gene Alias AI841080; Caspr; caspr1; CNTNAP; CNTNAP1; contactin associated protein 1; contactin associated protein-like 1; contactin-associated protein 1; LOW QUALITY PROTEIN: contactin-associated protein 1; MHDNIV; NCP1; neurexin 4; neurexin 4 (contactin associated protein); neurexin IV; Neurexin-4; NRXN4; p190; paranodin; shm
Gene Symbols Cntnap1
Host Species Rabbit
Immunogen A synthesized peptide derived from human Caspr (700-770AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53321, 8506
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less