missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNTF Polyclonal Antibody

Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA118365
Description
Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Avoid repetitive freeze/thaw cycles. Glycerol (1:1) may be added for an additional stability.
The protein encoded by this gene is a polypeptide hormone whose actions appear to be restricted to the nervous system where it promotes neurotransmitter synthesis and neurite outgrowth in certain neuronal populations. The protein is a potent survival factor for neurons and oligodendrocytes and may be relevant in reducing tissue destruction during inflammatory attacks. A mutation in this gene, which results in aberrant splicing, leads to ciliary neurotrophic factor deficiency, but this phenotype is not causally related to neurologic disease. A read-through transcript variant composed of ZFP91 and CNTF sequence has been identified, but it is thought to be non-coding. Read-through transcription of ZFP91 and CNTF has also been observed in mouse.Chemical Identifiers
| CNTF | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Polyclonal | |
| Lyophilized | |
| IgG | |
| Human, Mouse, Rat | |
| CNTF | |
| AI429687; Ciliary neurotrophic factor; ciliary neurotropic factor; Cntf; HCNTF; H-CNTF | |
| CNTF | |
| Primary |
| Conc. Not Determined | |
| ELISA, Western Blot | |
| Unconjugated | |
| Sheep | |
| RUO | |
| Whole serum with no preservative | |
| P20294, P26441, P51642 | |
| 1270, 12803, 25707 | |
| Recombinant human CNTF. | |
| Antibody |
Specifications
| CNTF | |
| Polyclonal | |
| Unconjugated | |
| CNTF | |
| AI429687; Ciliary neurotrophic factor; ciliary neurotropic factor; Cntf; HCNTF; H-CNTF | |
| Sheep | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Conc. Not Determined | |
| Whole serum with no preservative | |
| P20294, P26441, P51642 | |
| CNTF | |
| Recombinant human CNTF. | |
| RUO | |
| 1270, 12803, 25707 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |