missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CNOT7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa5101270
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101270 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101270 Supplier Invitrogen™ Supplier No. PA5101270
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CNOT7.

CNOT7 (CCR4-NOT transcription complex subunit 7) has 3'-5' poly(A) exoribonuclease activity for synthetic poly(A) RNA substrate. Its function seems to be partially redundant with that of CNOT8. CNOT7 is a catalytic component of the CCR4-NOT complex, which is one of the major cellular mRNA deadenylases, and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation, and general transcription regulation. During miRNA-mediated repression, the complex seems to act as a transcriptional repressor during translational initiation, as well. Additional complex functions may be a consequence of its influence on mRNA expression. CNOT7 associates with members of the BTG family such as TOB1 and BTG2 and is required for their anti-proliferative activity.

Specifications

Antigen CNOT7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CNOT7
Gene Accession No. Q60809, Q9UIV1
Gene Alias AU022737; BTG1-binding factor 1; CAF1; CAF-1; Caf1a; carbon catabolite repressor protein (CCR4)-associative factor 1; catabolite repressor protein (CCR4)-associative factor 1; CCR4-associated factor 1; CCR4-NOT transcription complex subunit 7; CCR4-NOT transcription complex subunit-like protein; CCR4-NOT transcription complex, subunit 7; CNOT7; fd59c07; fj07d05; hCAF-1; hypothetical protein LOC508055; Pop2; POP2 homolog; wu:fd59c07; wu:fj07d05; zgc:153168
Gene Symbols CNOT7
Host Species Rabbit
Immunogen A synthesized peptide derived from human CNOT7(Accession Q9UIV1), corresponding to amino acid residues E243-S285.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18983, 29883
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less