Learn More
Invitrogen™ CNOT4 Monoclonal Antibody (OTI3D11)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA525914
Catalog No. PIMA525914
Only null left
Explore available promotions
Description
Description
CNOT4 Monoclonal Antibody for Western Blot, Flow
CNOT4 is a 575 amino acid protein with E3 ubiquitin ligase activity, mainly involved in protein modification and protein ubiquitination. It contains a ring type zinc finger domain of the C4C4 type, an RNA recognition motif and a bipartite nuclear localization signal.Studies show that CNOT4 is a component of the CCR4-NOT (CAF1) core complex that functions as general transcription regulation complex and has both a positive as well as a negative effect on the transcription of multiple functionally unrelated genes, thus controls gene expression. CNOT4 interacts with CNOT1 via C-terminus and with E2 ubiquitin ligases via C4C4 RING domain. Reports suggest that CNOT4 gets phosphorylated in response to DNA damage on consensus sites recognized by ATM and ATR.
Specifications
Specifications
CNOT4
Monoclonal
0.81 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O95628
CNOT4
Human recombinant protein fragment corresponding to ammino acids 190-455 of CNOT4 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Western Blot
OTI3D11
Unconjugated
CNOT4
CCR4-associated factor 4; CCR4-NOT transcription complex subunit 4; CCR4-NOT transcription complex subunit 4 L homeolog; CCR4-NOT transcription complex subunit 4 S homeolog; CCR4-NOT transcription complex subunit 4 S homeolog; CCR4-NOT transcription complex, subunit 4; CCR4-NOT transcription complex, subunit 4 isoform a; CLONE243; cnot4; cnot4.S; E3 ubiquitin-protein ligase CNOT4; hypothetical protein; negative regulator of transcription 4; Not4; NOT4 (negative regulator of transcription 4, yeast) homolog; Not4h; Not4hp; Potential transcriptional repressor NOT4Hp; RING-type E3 ubiquitin transferase CNOT4; transcriptional repressor NOT4Hp; XELAEV_18019807mg
Mouse
Affinity Chromatography
RUO
4850
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions