missing translation for 'onlineSavingsMsg'
Learn More

Cytiva CNBr-Activated Sepharose™ 4 Fast Flow Affinity Media

Catalog No. 45002969
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002969 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002969 Supplier Cytiva Supplier No. 17098101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for coupling of large amino-containing ligands

 

  • Resin composed of cross-linked 4% agarose beads that have been pre-activated with cyanogen bromide
  • Proteins and other molecules containing primary amino groups are coupled directly to preactivated gel
  • Allows for multi-point attachment of protein ligands which minimizes ligand leakage
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

 

Specifications

Quantity 10 g
For Use With (Application) For Coupling of Large Amino-Containing Ligands
Stability Stable in commonly used aqueous Buffers provided that the ligand withstand the pH or the chemical environment
Storage Requirements 2°C to 8°C
Product Type CNBr-Activated Sepharose 4 Fast Flow Media
Flow Rate 150 to 250 cm/Hoμr
Particle Size 45 to 165 μm
Height (English) Bed 9.84 in.
Height (Metric) Bed 25 cm
Includes Certificate of Analysis
Max. Pressure 100 kPa
Matrix 4% Cross-Linked Agarose
pH Range 3 to 11
Storage Buffer 20% Ethanol
Ligand Type Cyanogen Bromide Activated
Show More Show Less