missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CMTM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582489
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582489 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582489 Supplier Invitrogen™ Supplier No. PA582489
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: NDVAKFLKQG DSADETTAHK TEEENSDSDS Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 83%.

This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies of signaling molecules. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 16. Alternatively spliced transcript variants containing different 5' UTRs, but encoding the same protein, have been identified.

Specifications

Antigen CMTM3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CMTM3
Gene Accession No. Q96MX0
Gene Alias 9430096L06Rik; AI413895; BNAS2; chemokine-like factor super family 3; chemokine-like factor superfamily member 3; CKLF like MARVEL transmembrane domain containing 3; CKLF-like MARVEL transmembrane domain containing 3; CKLF-like MARVEL transmembrane domain-containing protein 3; CKLFSF3; CMTM3
Gene Symbols CMTM3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CMTM3. Recombinant protein control fragment (Product #RP-90797).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123920
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less