missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ CMP-Sialic Acid, Disodium Salt, Calbiochem™,

Catalog No. 2332645MG
Change view
Click to view available options
Quantity:
5 mg
25 mg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2332645MG 5 mg Glass Bottle
23326425MG 25 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2332645MG Supplier MilliporeSigma™ Supplier No. 2332645MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recommended Storage

Stock solutions are Stable for up to 3 months at −20°C Unstable under acidic conditions.

Chemical Identifiers

CAS 3063-71-6
Molecular Formula C20H31N4O16P
Molecular Weight (g/mol) 614.454
InChI Key TXCIAUNLDRJGJZ-CEUYIXQXSA-N
Synonym CMP-N-Acetylneuraminic Acid, 2Na, CMP-NANA, CMP-Neu5Ac
PubChem CID 131632558
IUPAC Name (2R,4S,5R,6R)-5-acetamido-2-[[(2R,3R,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-4-hydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid
SMILES CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)OP(=O)(O)OCC2C(C(C(O2)N3C=CC(=NC3=O)N)O)O)O

Specifications

Color White
Packaging Glass Bottle
Quantity 5 mg
Percent Purity ≥95% by HPLC
Recommended Storage Stock solutions are Stable for up to 3 months at −20°C Unstable under acidic conditions.