accessibility menu, dialog, popup

UserName

CAS RN 3063-71-6

IUPAC Name: 2-[({[5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy]-5-acetamido-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Synonyms: 2-({[5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy(hydroxy)phosphoryl}oxy)-5-acetamido-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Molecular Weight (g/mol): 614.45
Molecular Formula: C20H31N4O16P
InChi Key: TXCIAUNLDRJGJZ-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(O)CC(OP(O)(=O)OCC2OC(C(O)C2O)N2C=CC(N)=NC2=O)(OC1C(O)C(O)CO)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results