missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CMG1 Polyclonal Antibody, Biotin

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
This gene encodes a core intraflagellar transport (IFT) protein which belongs to a multi-protein complex involved in the transport of ciliary proteins along axonemal microtubules. IFT proteins are found at the base of the cilium as well as inside the cilium, where they assemble into long arrays between the ciliary base and tip. This protein, together with intraflagellar transport protein 81, binds and transports tubulin within cilia and is required for ciliogenesis. Naturally occurring mutations in this gene are associated with amyotrophic lateral sclerosis--frontotemporal dementia and Bardet-Biedl Syndrome.
Specifications
Specifications
| Antigen | CMG1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Biotin |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | Ift74 |
| Gene Accession No. | Q96LB3 |
| Gene Alias | 1700029H06Rik; b2b796Clo; capillary morphogenesis gene 1 protein; capillary morphogenesis protein 1; CCDC2; CMG1; CMG-1; coiled-coil domain containing 2; coiled-coil domain-containing protein 2; Ift74; intraflagellar transport 74; intraflagellar transport 74 homolog; intraflagellar transport protein 74 homolog; LOW QUALITY PROTEIN: intraflagellar transport protein 74 homolog |
| Gene Symbols | Ift74 |
| Show More |