missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLPX Recombinant Rabbit Monoclonal Antibody (23GB1745)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA552591
Description
CLPX Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
ATP-dependent specificity component of the Clp protease. It directs the protease to specific substrates. Can perform chaperone functions in the absence of clpP.Chemical Identifiers
| CLPX | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| CLPX | |
| ATP-dependent Clp protease ATP-binding subunit clpX-like, mitochondrial; AU014732; caseinolytic mitochondrial matrix peptidase chaperone subunit; caseinolytic peptidase X; caseinolytic protease X; CLPX; ClpX caseinolytic peptidase X homolog; ClpX caseinolytic protease X homolog; E330029I21; energy-dependent regulator of proteolysis | |
| CLPX | |
| Primary | |
| Affinity chromatography |
| 23GB1745 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O76031, Q5U2U0, Q9JHS4 | |
| 10845, 270166, 300786 | |
| A synthesized peptide derived from human CLPX (600-633AA). | |
| Antibody |
Specifications
| CLPX | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CLPX | |
| ATP-dependent Clp protease ATP-binding subunit clpX-like, mitochondrial; AU014732; caseinolytic mitochondrial matrix peptidase chaperone subunit; caseinolytic peptidase X; caseinolytic protease X; CLPX; ClpX caseinolytic peptidase X homolog; ClpX caseinolytic protease X homolog; E330029I21; energy-dependent regulator of proteolysis | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10845, 270166, 300786 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1745 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O76031, Q5U2U0, Q9JHS4 | |
| CLPX | |
| A synthesized peptide derived from human CLPX (600-633AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |