missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CLPTM1 Recombinant Rabbit Monoclonal Antibody (23GB3090)
GREENER_CHOICE

Catalog No. MA553320
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553320 100 μL
MA553321 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553320 Supplier Invitrogen™ Supplier No. MA553320
Only null left

Rabbit Recombinant Monoclonal Antibody

CLPTM1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

CLPTM1 may play a role in T-cell development.

Specifications

Antigen CLPTM1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3090
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CLPTM1
Gene Accession No. O96005, Q8VBZ3
Gene Alias cleft lip and palate associated transmembrane protein 1; cleft lip and palate transmembrane protein 1; Cleft lip and palate transmembrane protein 1 homolog; CLPTM1; CLPTM1, transmembrane protein; HS9; N14; Thymic epithelial cell surface antigen
Gene Symbols CLPTM1
Host Species Rabbit
Immunogen A synthesized peptide derived from CLPTM1 (30-54AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1209, 292696, 56457
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less