missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Clotrimazole | |
| 147°C | |
| C22H17ClN2 | |
| 25 g | |
| VNFPBHJOKIVQEB-UHFFFAOYSA-N | |
| 1-[(2-chlorophenyl)-diphenylmethyl]imidazole | |
| 2812 | |
| 344.84 | |
| Crystalline Powder |
| 23593-75-1 | |
| White | |
| MFCD00057220 | |
| clotrimazole, lotrimin, canesten, mycelex, mycosporin, empecid, clotrimazol, mykosporin, gyne lotrimin, chlotrimazole | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3Cl)N4C=CN=C4 | |
| 344.842 | |
| CHEBI:3764 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 23593-75-1 | |
| 344.842 | |
| VNFPBHJOKIVQEB-UHFFFAOYSA-N | |
| 2812 | |
| 1-[(2-chlorophenyl)-diphenylmethyl]imidazole |
| C22H17ClN2 | |
| MFCD00057220 | |
| clotrimazole, lotrimin, canesten, mycelex, mycosporin, empecid, clotrimazol, mykosporin, gyne lotrimin, chlotrimazole | |
| CHEBI:3764 | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3Cl)N4C=CN=C4 |
Safety and Handling
RTECSNumber : NI4377000
TSCA : No