missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Clotrimazole

Catalog No. AC455460250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455460250 25 g
AC455461000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455460250 Supplier Thermo Scientific Chemicals Supplier No. 455460250
Only null left

CAS: 23593-75-1 | C22H17ClN2 | 344.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23593-75-1
Molecular Formula C22H17ClN2
Molecular Weight (g/mol) 344.84
InChI Key VNFPBHJOKIVQEB-UHFFFAOYSA-N
Synonym clotrimazole, lotrimin, canesten, mycelex, mycosporin, empecid, clotrimazol, mykosporin, gyne lotrimin, chlotrimazole
PubChem CID 2812
ChEBI CHEBI:3764
IUPAC Name 1-[(2-chlorophenyl)-diphenylmethyl]imidazole
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3Cl)N4C=CN=C4

Specifications

Melting Point 141.0°C to 145.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Quantity 25 g
Formula Weight 344.84
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Clotrimazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
EINECSNumber 245-764-8

RUO – Research Use Only