missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Closures for Snap Ring Vials with Snap Cap
Closure type clear polyethylene snap cap
Specifications
| Description | Snap cap |
|---|---|
| Color | Red, Clear |
| For Use With (Application) | 11 mm Snap Ring |
| No. per Pack | 100 |
| Catalog Number | Mfr. No. | Material | Hardness | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | Hardness | Price | Quantity & Availability | ||||
|
111011911
|
MilliporeSigma™ Supelco™
SU860093 |
Polyethylene, PTFE, Silicone | 45 (Durometer: shore A) |
N/A
|
|
||||
Description
- Closure type clear polyethylene snap cap
- Compatible with Large Opening Crimp/Snap Seal Vials
- Clear polyethylene cap
Specifications
| Snap cap | |
| Red, Clear | |
| 11 mm Snap Ring | |
| 100 |