missing translation for 'onlineSavingsMsg'
Learn More

Clopidogrel Related Compound A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013121
Change view
Click to view available options
Quantity:
40 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013121 40 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013121 Supplier Merck Emd Millipore Supplier No. PHR199040MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 144750-42-5
Linear Formula C15H14 ClNO2 S · HCl
MDL Number MFCD08063656
Synonym -(2-Chlorophenyl)-6,7-dihydrothieno[3,2-c]pyridine-5(4 H)-acetic acid hydrochloride
Type Certified Reference Material
Quantity 40 mg
Formula Weight 344.26
Grade Certified Reference Material