missing translation for 'onlineSavingsMsg'
Learn More

Clomipramine hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018858
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018858 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018858 Supplier Merck Emd Millipore Supplier No. PHR17201G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Clomipramine hydrochloride is one of the dibenzazepine tricyclic antidepressants that inhibits noradrenalin and serotonin reuptake in central nervous system. It also shows antimuscarinic properties. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 17321-77-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C19H23 ClN2 · HCl
MDL Number MFCD00069234
Synonym Clomipramine hydrochloride; 3-Chloro-10,11-dihydro-N,N-dimethyl-5 H-dibenz[b,f]azepine-5-propanamine hydrochloride; Anafranil hydrochloride
Type Certified Reference Material
Quantity 1 g
Formula Weight 351.31
Grade Certified Reference Material