missing translation for 'onlineSavingsMsg'
Learn More

Clomiphene citrate

Catalog No. p-7042757
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6366304 2 g
AAJ6366309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6366304 Supplier Thermo Scientific Chemicals Supplier No. J6366304

A selective estrogen receptor modulator | CAS: 50-41-9 | C32H36ClNO8 | 598.089 g/mol

Selective estrogen receptor modulator, which acts by inhibiting the action of estrogen on the pituitary. Clomiphene Citrate is an activator of PKC and Gonad-stimulating principle.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Selective estrogen receptor modulator, which acts by inhibiting the action of estrogen on the pituitary. Clomiphene Citrate is an activator of PKC and Gonad-stimulating principle.

Solubility
Slightly soluble in water, methanol, chloroform (slightly), and DMSO (10 mM). Insoluble in ether.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 50-41-9
Molecular Formula C32H36ClNO8
Molecular Weight (g/mol) 598.089
MDL Number MFCD00058322
InChI Key PYTMYKVIJXPNBD-BTKVJIOYSA-N
Synonym clomifene citrate, clomiphene citrate, clomid, cis-clomiphene citrate, clomiphene a citrate, zuclomid, chloramiphene, milophene, serophene, z-clomiphene citrate
PubChem CID 6420009
IUPAC Name 2-[4-[(E)-2-chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CCN(CC)CCOC1=CC=C(C=C1)C(=C(C2=CC=CC=C2)Cl)C3=CC=CC=C3.C(C(=O)O)C(CC(=O)O)(C(=O)O)O

Specifications

Color White
Quantity 2 g
Merck Index 14,2386
Solubility Information Slightly soluble in water,methanol,chloroform (slightly),and DMSO (10 mM). Insoluble in ether.
Formula Weight 598.08
Physical Form Crystalline Powder
Chemical Name or Material Clomiphene citrate
Hazard Category H360
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 200-035-3
RTECSNumber YE0875000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only