missing translation for 'onlineSavingsMsg'
Learn More

Clofibrate 98.0+%, TCI America™

Catalog No. p-7124696
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C094125G 25 g
C0941500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C094125G Supplier TCI America Supplier No. C094125G
Only null left

Chemical Identifiers

CAS 637-07-0
Molecular Formula C12H15ClO3
Molecular Weight (g/mol) 242.70
MDL Number MFCD00000615
InChI Key KNHUKKLJHYUCFP-UHFFFAOYSA-N
Synonym clofibrate, ethyl clofibrate, clofibratum, clofibrato, atromid-s, miscleron, atromid, liprin, epib, angiokapsul
PubChem CID 2796
ChEBI CHEBI:3750
IUPAC Name ethyl 2-(4-chlorophenoxy)-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1

Specifications

Color Yellow
Boiling Point 150°C
Quantity 25 g
Formula Weight 242.70
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Clofibrate
EINECSNumber (9)-0266
RTECSNumber UE9480000
TSCA No