missing translation for 'onlineSavingsMsg'
Learn More

Clofibrate, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019520
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019520 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019520 Supplier Merck Emd Millipore Supplier No. 41326100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified analytical standard intended for use in forensics and toxicology applications.

Pharmaceutical (small molecule)

Specifications

Chemical Name or Material Clofibrate
CAS 637-07-0
Flash Point 113°C
Physical Form Neat
Linear Formula C12H15 ClO3
Technique Liquid Chromatography
MDL Number MFCD00000615
Synonym 2-(4-Chlorophenoxy)-2-methylpropionic acid ethyl ester; Ethyl 2-(4-chlorophenoxy)-2-methylpropionate; Ethyl 2-(4-chlorophenoxy)isobutyrate
Type Analytical Standard
Molecular Weight (g/mol) 242.70
Quantity 100 mg
Formula Weight 242.70
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less