Learn More
Clofibrate, 95+%
CAS: 637-07-0 | C12H15ClO3 | 242.70 g/mol
Supplier: Thermo Scientific Chemicals J6034214
Description
It is a lipid-lowering agent used for controlling the high cholesterol and triacylglyceride level in the blood. It increases lipoprotein lipase activity to promote the conversion of VLDL to LDL, and hence reduce the level of VLDL. It can increase the level of HDL as well. Clofibrate is a ligand agonist for PPARα (peroxisome proliferator-activated receptor α), or an inducer of cytochromes P450âs CYP4A1 and CYP4A3.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a lipid-lowering agent used for controlling the high cholesterol and triacylglyceride level in the blood. It increases lipoprotein lipase activity to promote the conversion of VLDL to LDL, and hence reduce the level of VLDL. It can increase the level of HDL as well. Clofibrate is a ligand agonist for PPARα (peroxisome proliferator-activated receptor α), or an inducer of cytochromes P450âs CYP4A1 and CYP4A3.
Solubility
Soluble to 100 mM in DMSO.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat, bases. Store in refrigerator.
Specifications
| Clofibrate | |
| 1.14 | |
| 113°C (235°F) | |
| 1.5 | |
| 25 g | |
| 1913459 | |
| clofibrate, ethyl clofibrate, clofibratum, clofibrato, atromid-s, miscleron, atromid, liprin, epib, angiokapsul | |
| KNHUKKLJHYUCFP-UHFFFAOYSA-N | |
| ethyl 2-(4-chlorophenoxy)-2-methylpropanoate | |
| 2796 | |
| 242.7 | |
| Liquid |
| 637-07-0 | |
| 154°C to 156°C | |
| C12H15ClO3 | |
| MFCD00000615 | |
| UN3082 | |
| 14,2377 | |
| Soluble to 100 mM in DMSO. | |
| CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1 | |
| 242.70 | |
| CHEBI:3750 | |
| ≥95% |
Chemical Identifiers
| 637-07-0 | |
| 242.70 | |
| KNHUKKLJHYUCFP-UHFFFAOYSA-N | |
| 2796 | |
| ethyl 2-(4-chlorophenoxy)-2-methylpropanoate |
| C12H15ClO3 | |
| MFCD00000615 | |
| clofibrate, ethyl clofibrate, clofibratum, clofibrato, atromid-s, miscleron, atromid, liprin, epib, angiokapsul | |
| CHEBI:3750 | |
| CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1 |
Safety and Handling
GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 211-277-4
RTECSNumber : UE9480000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only