missing translation for 'onlineSavingsMsg'
Learn More

Clofibrate, 95+%

CAS: 637-07-0 | C12H15ClO3 | 242.70 g/mol

Supplier:  Thermo Scientific Chemicals J6034206

 View more versions of this product

Catalog No. AAJ6034206

Only null left
Explore available promotions

Description

Description

It is a lipid-lowering agent used for controlling the high cholesterol and triacylglyceride level in the blood. It increases lipoprotein lipase activity to promote the conversion of VLDL to LDL, and hence reduce the level of VLDL. It can increase the level of HDL as well. Clofibrate is a ligand agonist for PPARα (peroxisome proliferator-activated receptor α), or an inducer of cytochromes P450’s CYP4A1 and CYP4A3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble to 100 mM in DMSO.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat, bases. Store in refrigerator.
Specifications

Specifications

Clofibrate
1.14
113°C (235°F)
1.5
5 g
1913459
clofibrate, ethyl clofibrate, clofibratum, clofibrato, atromid-s, miscleron, atromid, liprin, epib, angiokapsul
KNHUKKLJHYUCFP-UHFFFAOYSA-N
ethyl 2-(4-chlorophenoxy)-2-methylpropanoate
2796
242.7
Liquid
637-07-0
154°C to 156°C
C12H15ClO3
MFCD00000615
UN3082
14,2377
Soluble to 100 mM in DMSO.
CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1
242.70
CHEBI:3750
≥95%

Chemical Identifiers

637-07-0
242.70
KNHUKKLJHYUCFP-UHFFFAOYSA-N
2796
ethyl 2-(4-chlorophenoxy)-2-methylpropanoate
C12H15ClO3
MFCD00000615
clofibrate, ethyl clofibrate, clofibratum, clofibrato, atromid-s, miscleron, atromid, liprin, epib, angiokapsul
CHEBI:3750
CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H315-H318-H335

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 211-277-4

RTECSNumber : UE9480000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only