missing translation for 'onlineSavingsMsg'
Learn More

Clofarabine 98.0+%, TCI America™

Supplier:  TCI America C2500100MG

Quantity 100 mg

 View more versions of this product

Catalog No. C2500100MG


Only null left

Description

Description

Specifications

Chemical Identifiers

123318-82-1
285.69
ADXUXRNLBYKGOA-GPGUAWMRNA-N
119182
(2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-4-chloro-2-(hydroxymethyl)oxolan-3-ol
C10H12ClN5O3
MFCD00871077
clofarabine, clolar, evoltra, clofarex, cafda, clofarabina, clofarabinum, clofarabine usan, cl-f-ara-a, c1-f-ara-a
CHEBI:681569
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3Cl)C2=NC=N1

Specifications

Clofarabine
227°C
C10H12ClN5O3
MFCD00871077
ADXUXRNLBYKGOA-GPGUAWMRNA-N
(2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-4-chloro-2-(hydroxymethyl)oxolan-3-ol
119182
303.68
Crystalline Powder
123318-82-1
White
100 mg
clofarabine, clolar, evoltra, clofarex, cafda, clofarabina, clofarabinum, clofarabine usan, cl-f-ara-a, c1-f-ara-a
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3Cl)C2=NC=N1
285.69
CHEBI:681569
≥98.0% (HPLC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UD7473000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions