accessibility menu, dialog, popup

UserName

CAS RN 123318-82-1

IUPAC Name: (2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-4-chloro-2-(hydroxymethyl)oxolan-3-ol
Synonyms: (2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-4-chloro-2-(hydroxymethyl)oxolan-3-ol
Molecular Weight (g/mol): 285.69
Molecular Formula: C10H12ClN5O3
InChi Key: ADXUXRNLBYKGOA-GPGUAWMRNA-N
SMILES: NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3Cl)C2=NC=N1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)